2,5-Dichlorophenyl isocyanate 97%

Leverancier: Apollo Scientific

APOSOR015004-25GEA 256 EUR
APOSOR015004-25G APOSOR015004-5G
2,5-Dichlorophenyl isocyanate 97%
2,5-Dichlorophenyl isocyanate

  • May form small amount of sediment.

Formule: C₇H₃Cl₂NO
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00002002
CAS-nummer: 5392-82-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR