2,3-Dimethylindole 97%
Leverancier: Apollo Scientific
|
Waarschuwing
|
APOSOR10173-25GEA
91.6
EUR
APOSOR10173-25G
2,3-Dimethylindole 97%
2,3-Dimethylindole
Formule:
C₁₀H₁₁N MW: 145,2 g/mol Kookpunt: 285 °C Smeltpunt: 105…107 °C Opslagtemperatuur: Koelkast |
MDL-nummer:
MFCD00005617 CAS-nummer: 91-55-4 EINECS: 202-076-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...