N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride 98%

Waarschuwing

APOSOR11243-250GEA 0 EUR
APOSOR11243-250G APOSOR11243-50G
N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride 98%
N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride
Formule: C₁₀H₁₈Cl₂N₂
MW: 237,17 g/mol
Kookpunt: 222…224 °C
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00012482
CAS-nummer: 637-01-4
EINECS: 211-274-8

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR