6-Nitro-2-picoline 95+%

APOSOR11388-5GEA 0 EUR
APOSOR11388-5G APOSOR11388-1G APOSOR11388-10G
6-Nitro-2-picoline 95+%
6-Nitro-2-picoline
Formule: C₆H₆N₂O₂
MW: 138,1240 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD04114203
CAS-nummer: 18368-61-1

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR