2-(Bromomethyl)-4-chlorophenylboronic acid pinacol ester 95%

APOSOR11450-250MGEA 0 EUR
APOSOR11450-250MG APOSOR11450-1G
2-(Bromomethyl)-4-chlorophenylboronic acid pinacol ester 95%
2-(Bromomethyl)-4-chlorophenylboronic acid pinacol ester
Formule: C₁₃H₁₇BBrClO₂
MW: 331,4409 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD09475815
CAS-nummer: 957034-64-9

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR