4-(2-Bromophenylsulphonyl)morpholine

Waarschuwing

APOSOR11692-25GEA 0 EUR
APOSOR11692-25G APOSOR11692-5G
4-(2-Bromophenylsulphonyl)morpholine
4-(2-Bromophenylsulphonyl)morpholine
Formule: C₁₀H₁₂BrNO₃S
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD09340792
CAS-nummer: 688798-57-4

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR