N-(4-Methoxybenzyl) 5-bromopicolinamide

Leverancier: Apollo Scientific

Waarschuwing

APOSOR11923-250MGEA 58.2 EUR
APOSOR11923-250MG APOSOR11923-1G
N-(4-Methoxybenzyl) 5-bromopicolinamide
N-(4-Methoxybenzyl) 5-bromopicolinamide
Formule: C₁₄H₁₃BrN₂O₂
MW: 321,1692 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD09801060
CAS-nummer: 951885-02-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR