4,6-Dichloropyrimidine-5-carbaldehyde 97%
|
Waarschuwing
|
APOSOR14207-5GEA
0
EUR
APOSOR14207-5G
APOSOR14207-100G
APOSOR14207-25G
4,6-Dichloropyrimidine-5-carbaldehyde 97%
4,6-Dichloropyrimidine-5-carbaldehyde
Formule:
C₅H₂Cl₂N₂O MW: 175,99 g/mol Smeltpunt: 66…71 °C Opslagtemperatuur: Koelkast |
MDL-nummer:
MFCD02257701 CAS-nummer: 5305-40-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...