4-Cyclohexylphenylglyoxal hydrate 95%
Leverancier: Apollo Scientific
|
Waarschuwing
|
APOSOR1576-1GEA
0
EUR
APOSOR1576-1G
APOSOR1576-10G
APOSOR1576-25G
4-Cyclohexylphenylglyoxal hydrate 95%
4-Cyclohexylphenylglyoxal hydrate
Formule:
C₁₄H₁₈O₃ Opslagtemperatuur: Kamertemperatuur |
CAS-nummer:
99433-89-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...