3-Bromophenylglyoxal hydrate 95%

APOSOR1624-1GEA 0 EUR
APOSOR1624-1G
3-Bromophenylglyoxal hydrate 95%
3-Bromophenylglyoxal hydrate
Formule: C8H7BrO3
Opslagtemperatuur: Kamertemperatuur
CAS-nummer: 106134-16-1

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR