tert-Butyl-3-formyl-5-methoxy-1H-indole-1-carboxylate 98%

Leverancier: Apollo Scientific

APOSOR1659-1GEA 130 EUR
APOSOR1659-1G APOSOR1659-25G APOSOR1659-5G
tert-Butyl-3-formyl-5-methoxy-1H-indole-1-carboxylate 98%
tert-Butyl-3-formyl-5-methoxy-1H-indole-1-carboxylate
Formule: C₁₅H₁₇NO₄
MW: 275,2998 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD05864693
CAS-nummer: 324756-80-1

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR