(2-Benzyloxyphenyl)acetic acid
Leverancier: Apollo Scientific
|
Gevaar
|
APOSOR1661-5GEA
0
EUR
APOSOR1661-5G
APOSOR1661-1G
(2-Benzyloxyphenyl)acetic acid
(2-Benzyloxyphenyl)acetic acid
Formule:
C₆H₅CH₂OC₆H₄CH₂COOH MW: 242,27 g/mol Smeltpunt: 96…98 °C Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00016822 CAS-nummer: 22047-88-7 UN: 3077 ADR: 9+EHS,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...