1-(2-(4-Bromophenoxy)ethyl)-1H-pyrazole

APOSOR17038-5GEA 0 EUR
APOSOR17038-5G APOSOR17038-1G
1-(2-(4-Bromophenoxy)ethyl)-1H-pyrazole
1-(2-(4-Bromophenoxy)ethyl)-1H-pyrazole
Formule: C₁₁H₁₁BrN₂O
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD11855961
CAS-nummer: 1150271-30-9

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR