1H-Benzimidazole-5-boronic acid pinacol ester 95+%
Leverancier: Apollo Scientific
|
Waarschuwing
|
APOSOR17711-1GEA
94.5
EUR
APOSOR17711-1G
1H-Benzimidazole-5-boronic acid pinacol ester 95+%
1H-Benzimidazole-5-boronic acid pinacol ester
Formule:
C₁₃H₁₇BN₂O₂ MW: 244,10 g/mol Opslagtemperatuur: Vriezer |
MDL-nummer:
MFCD11054041 CAS-nummer: 1007206-54-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...