2-Ethynylbenzonitrile 99%
Leverancier: Apollo Scientific
|
Waarschuwing
|
APOSOR17876-250MGEA
14.7
EUR
APOSOR17876-250MG
APOSOR17876-1G
APOSOR17876-5G
2-Ethynylbenzonitrile 99%
2-Ethynylbenzonitrile
Formule:
C₉H₅N MW: 127,14 g/mol Smeltpunt: 75…76 °C Opslagtemperatuur: Vriezer |
MDL-nummer:
MFCD08703475 CAS-nummer: 40888-26-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...