3-Amino-4,5-dimethylisoxazole 97%

Leverancier: Apollo Scientific
Waarschuwing

APOSOR19546-1GEA 77.1 EUR
APOSOR19546-1G
3-Amino-4,5-dimethylisoxazole 97%
3-Amino-4,5-dimethylisoxazole
Formule: C5H8N2O
MW: 112,1300 g/mol
Smeltpunt: 114…116 °C
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD03411577
CAS-nummer: 13999-39-8

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR