3-Methyl-5-phenylisoxazole-4-carboxylic acid 97%
Leverancier: Apollo Scientific
|
Waarschuwing
|
APOSOR23335-5GEA
187
EUR
APOSOR23335-5G
APOSOR23335-10G
3-Methyl-5-phenylisoxazole-4-carboxylic acid 97%
3-Methyl-5-phenylisoxazole-4-carboxylic acid
Formule:
C11H9NO3 MW: 203,1941 g/mol Smeltpunt: 190…191 °C Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD03086120 CAS-nummer: 17153-21-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...