6-Chloro-m-toluic acid

Leverancier: Apollo Scientific
Waarschuwing

APOSOR2875-1GEA 12.6 EUR
APOSOR2875-1G
6-Chloro-m-toluic acid
6-Chloro-m-toluic acid
Formule: C₈H₇ClO₂
MW: 170,6 g/mol
Smeltpunt: 148…155 °C
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00045798
CAS-nummer: 6342-60-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR