2-Ethylimidazool 97%

Leverancier: Apollo Scientific

APOSOR2906-500GEA 125 EUR
APOSOR2906-500G APOSOR2906-1KG APOSOR2906-100G
2-Ethylimidazool 97%
2-Ethylimidazool
Formule: C₅H₈N₂
MW: 96,13 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00005192
CAS-nummer: 1072-62-4

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR