Methyl-1,2,5-trimethylpyrrole-3-carboxylate
APOSOR29450-5GEA
0
EUR
APOSOR29450-5G
APOSOR29450-1G
Methyl-1,2,5-trimethylpyrrole-3-carboxylate
Methyl-1,2,5-trimethylpyrrole-3-carboxylate
Formule:
C₉H₁₃NO₂ MW: 167,21 g/mol Smeltpunt: 110…115 °C Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00203867 CAS-nummer: 14186-50-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...