1-Methylisatin 97+%

Gevaar

APOSOR3000-5GEA 0 EUR
APOSOR3000-5G APOSOR3000-1G
1-Methylisatin 97+%
1-Methylisatin
Formule: C₉H₇NO₂
MW: 161,16 g/mol
Smeltpunt: 130…133 °C
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD00005812
CAS-nummer: 2058-74-4
EINECS: 218-164-9
UN: 2811
ADR: 6.1,III

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR