(4-Methylmorpholin-2-yl)methanamine

Leverancier: Apollo Scientific

APOSOR300162-1GEA 190 EUR
APOSOR300162-1G
(4-Methylmorpholin-2-yl)methanamine
(4-Methylmorpholin-2-yl)methanamine
Formule: C₆H₁₄N₂O
MW: 130,1882 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD03990582
CAS-nummer: 141814-57-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR