4-Chloro-2,3-dihydroinden-1-one 97%

Leverancier: Apollo Scientific

APOSOR301320-5GEA 69.8 EUR
APOSOR301320-5G
4-Chloro-2,3-dihydroinden-1-one 97%
4-Chloro-2,3-dihydroinden-1-one
Formule: C₉H₇ClO
MW: 166,6000 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD08544408
CAS-nummer: 15115-59-0

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR