Methyl-5-amino-2-methylbenzoate 97%
Leverancier: Apollo Scientific
APOSOR304201-25GEA
78.6
EUR
APOSOR304201-25G
Methyl-5-amino-2-methylbenzoate 97%
Methyl-5-amino-2-methylbenzoate
Formule:
C₉H₁₁NO₂ MW: 165,19 g/mol Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD08752568 CAS-nummer: 18595-12-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...