Methyl benzo[d]thiazole-7-carboxylate

Leverancier: Apollo Scientific

APOSOR305041-250MGEA 121 EUR
APOSOR305041-250MG
Methyl benzo[d]thiazole-7-carboxylate
Methyl benzo[d]thiazole-7-carboxylate
Formule: C₉H₇NO₂S
MW: 193,22 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD13190218
CAS-nummer: 1038509-28-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR