4-Amino-6-methylquinoline

Leverancier: Apollo Scientific

APOSOR307946-1GEA 376 EUR
APOSOR307946-1G
4-Amino-6-methylquinoline
4-Amino-6-methylquinoline
Formule: C₁₀H₁₀N₂
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD06761164
CAS-nummer: 874589-77-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR