1-Methylazetidine-3-carboxylic acid 97%

Leverancier: Apollo Scientific
Waarschuwing

APOSOR308026-250MGEA 26.3 EUR
APOSOR308026-250MG APOSOR308026-1G
1-Methylazetidine-3-carboxylic acid 97%
1-Methylazetidine-3-carboxylic acid
Formule: C₅H₉NO₂
MW: 115,1300 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD11520621
CAS-nummer: 875629-26-8

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR