3-Isopropylmorpholine
Leverancier: Apollo Scientific
|
Waarschuwing
|
APOSOR308180-1GEA
241
EUR
APOSOR308180-1G
3-Isopropylmorpholine
3-Isopropylmorpholine
Formule:
C7H15NO MW: 129,2 g/mol Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD09054775 CAS-nummer: 927802-40-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...