4-Bromophenyl isothiocyanate
|
Waarschuwing
|
APOSOR3082-25GEA
0
EUR
APOSOR3082-25G
4-Bromophenyl isothiocyanate
4-Bromophenyl isothiocyanate
Formule:
C₇H₄BrNS MW: 214,09 g/mol Smeltpunt: 60…61 °C Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00004808 CAS-nummer: 1985-12-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...