3-Methylazetidine hydrochloride 95%

Leverancier: Apollo Scientific

APOSOR309000-1GEA 80 EUR
APOSOR309000-1G
3-Methylazetidine hydrochloride 95%
3-Methylazetidine hydrochloride
Formule: C4H10ClN
MW: 107,5800 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD09743448
CAS-nummer: 935669-28-6

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR