2-Methylquinolin-3-amine

Leverancier: Apollo Scientific

APOSOR309366-1GEA 390 EUR
APOSOR309366-1G
2-Methylquinolin-3-amine
2-Methylquinolin-3-amine
Formule: C₁₀H₁₀N₂
MW: 158,1998 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD08705651
CAS-nummer: 21352-22-7

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR