5-Aminovaleric acid

Leverancier: Apollo Scientific

APOSOR310635-5GEA 87.7 EUR
APOSOR310635-5G
5-Aminovaleric acid
5-Aminovaleric acid
Formule: C₅H₁₁NO₂
MW: 117,15 g/mol
Smeltpunt: 158…161 °C
Opslagtemperatuur: Kamertemperatuur
CAS-nummer: 660-88-8
EINECS: 211-544-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR