4-Iodo-2-methoxypyridine 98%

Leverancier: Apollo Scientific

APOSOR315216-1GEA 85.8 EUR
APOSOR315216-1G
4-Iodo-2-methoxypyridine 98%
4-Iodo-2-methoxypyridine
Formule: C₆H₆INO
MW: 235,0224 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD08277272
CAS-nummer: 98197-72-9

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR