(R)-N-Boc-piperidine-2-methanol 97%

APOSOR321384-5GEA 0 EUR
APOSOR321384-5G APOSOR321384-25G
(R)-N-Boc-piperidine-2-methanol 97%
(R)-N-Boc-piperidine-2-methanol
Formule: C₁₁H₂₁NO₃
MW: 215,2900 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD04973120
CAS-nummer: 134441-61-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR