1-Methyl-4-(4-nitrobenzyl)piperazine

Leverancier: Apollo Scientific

APOSOR322481-1GEA 184 EUR
APOSOR322481-1G
1-Methyl-4-(4-nitrobenzyl)piperazine
1-Methyl-4-(4-nitrobenzyl)piperazine
Formule: C₁₂H₁₇N₃O₂
Opslagtemperatuur: Kamertemperatuur
CAS-nummer: 70261-81-3

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR