5-Nitro-3-picoline 96%

APOSOR3238-5GEA 0 EUR
APOSOR3238-5G APOSOR3238-1G
5-Nitro-3-picoline 96%
5-Nitro-3-picoline
Formule: C₆H₆N₂O₂
MW: 138,13 g/mol
Smeltpunt: 93 °C
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD04114214
CAS-nummer: 6960-20-9

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR