6-Chloro-2-hydroxypyridine

Leverancier: Apollo Scientific

APOSOR345034-500GEA 536 EUR
APOSOR345034-500G APOSOR345034-100G APOSOR345034-25G
6-Chloro-2-hydroxypyridine
6-Chloro-2-hydroxypyridine
Formule: C₅H₄ClNO
MW: 129,55 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00006278
CAS-nummer: 16879-02-0

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR