6-Amino-3-bromopicolinic acid 98%

Leverancier: Apollo Scientific

APOSOR345173-1GEA 91.6 EUR
APOSOR345173-1G
6-Amino-3-bromopicolinic acid 98%
6-Amino-3-bromopicolinic acid
Formule: C₆H₅BrN₂O₂
MW: 217,0200 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD10699642
CAS-nummer: 1033201-61-4

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR