2-Bromo-3-chloro-5-hydroxypyridine 97%

APOSOR345246-1GEA 0 EUR
APOSOR345246-1G APOSOR345246-250MG APOSOR345246-5G
2-Bromo-3-chloro-5-hydroxypyridine 97%
2-Bromo-3-chloro-5-hydroxypyridine
Formule: C₅H₃BrClNO
MW: 208,4400 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD09835244
CAS-nummer: 130284-56-9

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR