2,6-Dichloro-3-methyl-5-nitropyridine 95+%
Leverancier: Apollo Scientific
APOSOR345475-1GEA
132
EUR
APOSOR345475-1G
2,6-Dichloro-3-methyl-5-nitropyridine 95+%
2,6-Dichloro-3-methyl-5-nitropyridine
Formule:
C₆H₄Cl₂N₂O₂ MW: 207,0100 g/mol Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD17014968 CAS-nummer: 58596-88-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...