4-(tert-Butyldimethylsilyloxy)-3-methoxyphenylboronic acid 98%

Leverancier: Apollo Scientific

Waarschuwing

APOSOR3485-1GEA 176 EUR
APOSOR3485-1G
4-(tert-Butyldimethylsilyloxy)-3-methoxyphenylboronic acid 98%
4-(tert-Butyldimethylsilyloxy)-3-methoxyphenylboronic acid
Formule: C₁₃H₂₃BO₄Si
Smeltpunt: 220…223 °C
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD09027250
CAS-nummer: 957060-93-4

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR