2-Ethylthiophenol

Leverancier: Apollo Scientific

APOSOR350160-25GEA 280 EUR
APOSOR350160-25G
2-Ethylthiophenol
2-Ethylthiophenol
Formule: C₈H₁₀S
MW: 138,23 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00010022
CAS-nummer: 4500-58-7

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR