Ethyl-4-methylpyrrole-2-carboxylate 95%

Leverancier: Apollo Scientific

APOSOR350171-1GEA 154 EUR
APOSOR350171-1G
Ethyl-4-methylpyrrole-2-carboxylate 95%
Ethyl-4-methylpyrrole-2-carboxylate
Formule: C₈H₁₁NO₂
MW: 153,1800 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00191668
CAS-nummer: 40611-85-6

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR