1-Methoxynaphthalene

Leverancier: Apollo Scientific

APOSOR350223-100GEA 61.1 EUR
APOSOR350223-100G
1-Methoxynaphthalene
1-Methoxynaphthalene
Formule: C₁₁H₁₀O
MW: 158,2 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00003924
CAS-nummer: 2216-69-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR