4-(2-Hydroxy-3-(piperidin-1-yl)propoxy)phenylboronic acid pinacol ester 98%

APOSOR3514-1GEA 0 EUR
APOSOR3514-1G
4-(2-Hydroxy-3-(piperidin-1-yl)propoxy)phenylboronic acid pinacol ester 98%
4-(2-Hydroxy-3-(piperidin-1-yl)propoxy)phenylboronic acid pinacol ester
Formule: C₂₀H₃₂BNO₄
Smeltpunt: 100…102 °C
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD09258654
CAS-nummer: 957061-08-4

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR