1-Boc-3-methyl-1H-pyrazole-5-boronic acid 95+%

Leverancier: Apollo Scientific

APOSOR360357-250MGEA 104 EUR
APOSOR360357-250MG
1-Boc-3-methyl-1H-pyrazole-5-boronic acid 95+%
1-Boc-3-methyl-1H-pyrazole-5-boronic acid
Formule: C₉H₁₅BN₂O₄
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD15143457
CAS-nummer: 1217501-27-3

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR