3-Oxo-3,4-dihydropyrazine-2-carboxylic acid 97%

Waarschuwing

APOSOR40047-25GEA 0 EUR
APOSOR40047-25G
3-Oxo-3,4-dihydropyrazine-2-carboxylic acid 97%
3-Oxo-3,4-dihydropyrazine-2-carboxylic acid
Formule: C5H4N2O3
MW: 140,0969 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00235136
CAS-nummer: 20737-42-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR