2-Bromo-6-chlorobenzoic acid 98%

Leverancier: Apollo Scientific

Gevaar

APOSOR40571-100GEA 459 EUR
APOSOR40571-100G
2-Bromo-6-chlorobenzoic acid 98%
2-Bromo-6-chlorobenzoic acid
Formule: C₇H₄BrClO₂
MW: 235,46 g/mol
Smeltpunt: 143…146 °C
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00672929
CAS-nummer: 93224-85-2
UN: 2811
ADR: 6.1,III

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR