3,5-Dichloro-4-nitropyridine

APOSOR41163-250MGEA 0 EUR
APOSOR41163-250MG APOSOR41163-1G APOSOR41163-5G
3,5-Dichloro-4-nitropyridine
3,5-Dichloro-4-nitropyridine
Formule: C₅H₂Cl₂N₂O₂
MW: 192,99 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD06254584
CAS-nummer: 433294-98-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR