Cyclopentylmethanol
Leverancier: Apollo Scientific
|
Waarschuwing
|
APOSOR46003-5GEA
14.7
EUR
APOSOR46003-5G
Cyclopentylmethanol
Cyclopentylmethanol
Formule:
C₆H₁₂O MW: 100,16 g/mol Kookpunt: 162…163 °C Dichtheid: 0,926 g/cm³ (25 °C) Vlampunt: 62 °C Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00001384 CAS-nummer: 3637-61-4 EINECS: 222-861-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...